eBioscience™ Streptavidin eFluor™ 570 Conjugate, 100 μg - FAQs
eBioscience™ Streptavidin eFluor™ 570 Conjugate, 100 μg - FAQs
View additional product information for eBioscience™ Streptavidin eFluor™ 570 Conjugate - FAQs (41-4317-82)