Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 570, eBioscience™ - FAQs
Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 570, eBioscience™ - FAQs (41-4321-82, 41-4321-80)