beta-3 Tubulin Monoclonal Antibody (2G10-TB3), eFluor™ 570, eBioscience™ - FAQs
beta-3 Tubulin Monoclonal Antibody (2G10-TB3), eFluor™ 570, eBioscience™ - FAQs
View additional product information for beta-3 Tubulin Monoclonal Antibody (2G10-TB3), eFluor™ 570, eBioscience™ - FAQs (41-4510-80)