Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 570, eBioscience™ - FAQs
Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 570, eBioscience™ - FAQs (41-4732-80)