Mouse anti-Rat IgG2b Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs
Mouse anti-Rat IgG2b Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs
View additional product information for Mouse anti-Rat IgG2b Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs (41-4815-82, 41-4815-80)