Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs
Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs
View additional product information for Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs (41-4817-80, 41-4817-82)