Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 570, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 570, eBioscience™ - FAQs (41-5699-82, 41-5699-80)