Mouse IgM Monoclonal Antibody (II/41), eFluor™ 570, eBioscience™ - FAQs
Mouse IgM Monoclonal Antibody (II/41), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Mouse IgM Monoclonal Antibody (II/41), eFluor™ 570, eBioscience™ - FAQs (41-5790-82, 41-5790-80)