SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), eFluor™ 570, eBioscience™ - FAQs
SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), eFluor™ 570, eBioscience™ - FAQs
View additional product information for SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), eFluor™ 570, eBioscience™ - FAQs (41-8843-82)