Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 570, eBioscience™ - FAQs
Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 570, eBioscience™ - FAQs (41-9003-80, 41-9003-82)