ERK1/2 Monoclonal Antibody (5AD13MA), eFluor™ 570, eBioscience™ - FAQs
ERK1/2 Monoclonal Antibody (5AD13MA), eFluor™ 570, eBioscience™ - FAQs
View additional product information for ERK1/2 Monoclonal Antibody (5AD13MA), eFluor™ 570, eBioscience™ - FAQs (41-9108-80, 41-9108-82)