AIRE Monoclonal Antibody (TM-724), eFluor™ 570, eBioscience™ - FAQs
AIRE Monoclonal Antibody (TM-724), eFluor™ 570, eBioscience™ - FAQs
View additional product information for AIRE Monoclonal Antibody (TM-724), eFluor™ 570, eBioscience™ - FAQs (41-9534-82)