Glucagon Monoclonal Antibody (ICACLS), eFluor™ 570, eBioscience™ - FAQs
Glucagon Monoclonal Antibody (ICACLS), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Glucagon Monoclonal Antibody (ICACLS), eFluor™ 570, eBioscience™ - FAQs (41-9743-82, 41-9743-80)