MAP2 Monoclonal Antibody (AP20), eFluor™ 570, eBioscience™ - FAQs
MAP2 Monoclonal Antibody (AP20), eFluor™ 570, eBioscience™ - FAQs
View additional product information for MAP2 Monoclonal Antibody (AP20), eFluor™ 570, eBioscience™ - FAQs (41-9763-82, 41-9763-80)