Collagen X Monoclonal Antibody (X53), eFluor™ 570, eBioscience™ - FAQs
Collagen X Monoclonal Antibody (X53), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Collagen X Monoclonal Antibody (X53), eFluor™ 570, eBioscience™ - FAQs (41-9771-80, 41-9771-82)