ZO-1 Monoclonal Antibody (R26.4C), eFluor™ 570, eBioscience™ - FAQs
ZO-1 Monoclonal Antibody (R26.4C), eFluor™ 570, eBioscience™ - FAQs
View additional product information for ZO-1 Monoclonal Antibody (R26.4C), eFluor™ 570, eBioscience™ - FAQs (41-9776-82, 41-9776-80)