Vinculin Monoclonal Antibody (7F9), eFluor™ 570, eBioscience™ - FAQs
Vinculin Monoclonal Antibody (7F9), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Vinculin Monoclonal Antibody (7F9), eFluor™ 570, eBioscience™ - FAQs (41-9777-82)