TRAF-1 Monoclonal Antibody (1F3), eFluor™ 570, eBioscience™ - FAQs
TRAF-1 Monoclonal Antibody (1F3), eFluor™ 570, eBioscience™ - FAQs
View additional product information for TRAF-1 Monoclonal Antibody (1F3), eFluor™ 570, eBioscience™ - FAQs (41-9791-82)