SOX2 Monoclonal Antibody (Btjce), eFluor™ 570, eBioscience™ - FAQs
SOX2 Monoclonal Antibody (Btjce), eFluor™ 570, eBioscience™ - FAQs
View additional product information for SOX2 Monoclonal Antibody (Btjce), eFluor™ 570, eBioscience™ - FAQs (41-9811-80)