Cytokeratin 18 Monoclonal Antibody (LDK18), eFluor™ 570, eBioscience™ - FAQs
Cytokeratin 18 Monoclonal Antibody (LDK18), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Cytokeratin 18 Monoclonal Antibody (LDK18), eFluor™ 570, eBioscience™ - FAQs (41-9815-82)