Nestin Monoclonal Antibody (10C2), eFluor™ 570, eBioscience™ - FAQs
Nestin Monoclonal Antibody (10C2), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Nestin Monoclonal Antibody (10C2), eFluor™ 570, eBioscience™ - FAQs (41-9843-80)