GFAP Monoclonal Antibody (GA5), eFluor™ 570, eBioscience™ - FAQs
GFAP Monoclonal Antibody (GA5), eFluor™ 570, eBioscience™ - FAQs
View additional product information for GFAP Monoclonal Antibody (GA5), eFluor™ 570, eBioscience™ - FAQs (41-9892-82, 41-9892-80)