Vimentin Monoclonal Antibody (V9), eFluor™ 570, eBioscience™ - FAQs
Vimentin Monoclonal Antibody (V9), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Vimentin Monoclonal Antibody (V9), eFluor™ 570, eBioscience™ - FAQs (41-9897-80, 41-9897-82)