Cytokeratin 19 Monoclonal Antibody (BA17), eFluor™ 570, eBioscience™ - FAQs
Cytokeratin 19 Monoclonal Antibody (BA17), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Cytokeratin 19 Monoclonal Antibody (BA17), eFluor™ 570, eBioscience™ - FAQs (41-9898-80)