Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0%, USP, GMP, Multi-Compendial, J.T.Baker™
Avantor™

Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0%, USP, GMP, Multi-Compendial, J.T.Baker™

This product is GMP Manufactured meets JPC (1997), BP, EP and USP Requirements.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량포장
4102-0712 kgPolyethylene Pail, Screw Top
4102-052.5 kgWhite Poly Bottle, Tamper-evident Cap
4102-F950 kgDrum, HDPE Flowmor Package
4102-0950 kgPolyethylene Drum, Open Head
4102-01500 gWhite Poly Bottle, Tamper-evident Cap
4102-F712 kgDrum, HDPE Flowmor Package
카탈로그 번호 4102-07
제품 가격(KRW)
-
수량:
12 kg
포장:
Polyethylene Pail, Screw Top
This product is GMP Manufactured meets JPC (1997), BP, EP and USP Requirements.
This product is GMP Manufactured meets JPC (1997), BP, EP and USP Requirements.
사양
끓는점357°C (1013 hPa)
CAS77-86-1
CAS77-86-1
화학물질 이름 또는 재질Tris(hydroxymethyl)aminomethane
색상Colorless
용융점169–172°C
분자식NH2C(CH2OH)3
포장Polyethylene Pail, Screw Top
동의어2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris(hydroxymethyl)methylamine, TRIS base
용도(애플리케이션)Pharma and Biopharma
등급USP
분자량(g/mol)121.14
백분율 순도99.0-101.0%
물리적 형태Crystalline
수량12 kg
용액 유형Buffer
pH10.0-11.5
Unit SizeEach