Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers
Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers
Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers
Thermo Scientific Chemicals

Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers

CAS: 928-55-2 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 410370100
Price (USD)
48.65
Special offer
Online exclusive
Ends: 28-Dec-2026
57.20
Save 8.55
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 28-Dec-2026
57.20
Save 8.55
Each
Chemical Identifiers
CAS928-55-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.3940 to 1.4000 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only