Ethylxanthic acid potassium salt, 97+%
Ethylxanthic acid potassium salt, 97+%
Ethylxanthic acid potassium salt, 97+%
Thermo Scientific Chemicals

Ethylxanthic acid potassium salt, 97+%

CAS: 140-89-6 | C3H5KOS2 | 160.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 410500250
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.00
Save 5.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
35.00
Save 5.35
Each
Chemical Identifiers
CAS140-89-6
IUPAC Namepotassium (ethoxymethanethioyl)sulfanide
Molecular FormulaC3H5KOS2
InChI KeyJCBJVAJGLKENNC-UHFFFAOYSA-M
SMILES[K+].CCOC([S-])=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to yellow
Infrared spectrumConforms
Loss on drying=<2 % (1 g, 105°C)
Appearance (Form)Powder or crystalline powder or crystals
Titration with HClO497.0 to 103.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only