Gallic acid, 98%
Gallic acid, 98%
Gallic acid, 98%
Thermo Scientific Chemicals

Gallic acid, 98%

CAS: 149-91-7 | C7H6O5 | 170.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 410860010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS149-91-7
IUPAC Name3,4,5-trihydroxybenzoic acid
Molecular FormulaC7H6O5
InChI KeyLNTHITQWFMADLM-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH97.5 to 102.5 %
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only