Geraniol, 99%
Geraniol, 99%
Geraniol, 99%
Thermo Scientific Chemicals

Geraniol, 99%

CAS: 106-24-1 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 410905000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-24-1
IUPAC Name(2E)-3,7-dimethylocta-2,6-dien-1-ol
Molecular FormulaC10H18O
InChI KeyGLZPCOQZEFWAFX-JXMROGBWSA-N
SMILESCC(C)=CCC\C(C)=C\CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4750 to 1.4780 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only