Hypersil™ PREP HS C8 Bulk Media
Actual product may vary
Thermo Scientific™
Hypersil™ PREP HS C8 Bulk Media
Ensure optimal chromatography performance and cost savings with Hypersil PREP HS C8 bulk chromatography media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
41110-10000B10000 g
41110-00050B50 g
41110-00100B100 g
41110-00500B500 g
41110-01000B1000 g
41110-05000B5000 g
Catalog number 41110-10000B
Price (USD)
-
Quantity:
10000 g
Ensure optimal chromatography performance with our Thermo Scientific™ Hypersil™ PREP HS C8 bulk chromatography media. Purchase bulk quantities of high surface area C8 media for faster separations in order to repack preparative HPLC columns yourself without paying for new hardware.
Repack Columns Yourself with Bulk Media
Similar selectivity, less retention than C18 columns.
  • Successful retention and separation of moderately hydrophobic analytes.
  • Ideal for fatty acids, which are too hydrophobic for analysis using C18 columns.
  • C8 (USP L7) selectivity.
  • High surface area particles with 100Å pore size.
  • pH stable from 2 to 8.
  • Maximum temperature of 60°C.
Faster Separations
  • Higher analytical throughput compared to C18 alkyl chain chemistry.
  • Quicker compound elution due to reduced hydrophobic interactions.
Customer Specific Orders
  • Please contact your local sales representative for Customer Specific Orders.
Specifications
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
PhaseReversed Phase
Pore Size100 Å
Product LinePrep LC
Quantity10000 g
Stationary PhaseC8
Surface Area (Metric)350 m²/g
Temperature60°C
USP TypeL7
Particle Size10 μm
pH2 to 8
Unit SizeEach