Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile
Thermo Scientific™

Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeClosure Type
4112-20002000 mL45-430 mmSolid, non-vented
4115-0125125 mL38-430 mmVented
4115-0250250 mL38-430 mmVented
4115-0500500 mL45-430 mmVented
4115-10001000 mL45-430 mmVented
4115-20002000 mL45-430 mmVented
4115-28002800 mL70 mmVented
4112-0125125 mL38-430 mmSolid, non-vented
4112-0250250 mL38-430 mmSolid, non-vented
4112-0500500 mL45-430 mmSolid, non-vented
4112-10001000 mL45-430 mmSolid, non-vented
4112-28002800 mL70 mmSolid, non-vented
Catalog number 4112-2000
Price (USD)
122.65
Online exclusive
138.00
Save 15.35
Case of 4
Capacity (Metric):
2000 mL
Closure Size:
45-430 mm
Closure Type:
Solid, non-vented
Price (USD)
122.65
Online exclusive
138.00
Save 15.35
Case of 4

Save preparation time and avoid contamination risk with Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features

  • Sterile and single use for reduced contamination risk
  • BPA-free
  • Collapses when autoclaved, reducing biohazardous waste
  • Graduated for quick volume assessment
  • Regulatory documentation available on request

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)64 oz.
Closure Size45-430 mm
Closure TypeSolid, non-vented
No. per Case4
No. per Pack1
ShapeErlenmeyer
Surface CoatingNone
AutoclavableNot Autoclavable
Bottom ShapePlain
Capacity (Metric)2000 mL
DisposableSingle-use
GraduatedYes
MaterialPETG
Mouth StyleNarrow
SterilitySterile
Unit SizeCase of 4