Hexadecyltrimethylammonium hydroxide, 25% in methanol
Hexadecyltrimethylammonium hydroxide, 25% in methanol
Hexadecyltrimethylammonium hydroxide, 25% in methanol
Thermo Scientific Chemicals

Hexadecyltrimethylammonium hydroxide, 25% in methanol

CAS: 505-86-2 | C19H43NO | 301.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
Packaging:
Glass bottle
Catalog number 411421000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS505-86-2
IUPAC Namehexadecyltrimethylazanium hydroxide
Molecular FormulaC19H43NO
InChI KeyWJLUBOLDZCQZEV-UHFFFAOYSA-M
SMILES[OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<3 % (K.F.)
Concentration23 to 27 w/w% (in methanol)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only