4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals
4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals
4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals

CAS: 3058-39-7 | Molecular Formula: C7H4IN | Molecular Weight: 229.02 g/mol
Have Questions?
Catalog NumberQuantity
4123800505 g
Catalog number 412380050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3058-39-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to yellow-brown
Infrared spectrumConforms
Melting point125°C to 128°C
GC>=98.5 %
Appearance (Form)Powder and/or chunks

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only