IR-125, pure, laser grade
IR-125, pure, laser grade
IR-125, pure, laser grade
Thermo Scientific Chemicals

IR-125, pure, laser grade

CAS: 3599-32-4 | C43H47N2NaO6S2 | 774.967 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
2 g
Packaging:
Glass bottle
Catalog number 412540020
Price (USD)
2,156.00
Each
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
2,156.00
Each
Chemical Identifiers
CAS3599-32-4
IUPAC Namesodium 2-[(5E)-7-[(2E)-1,1-dimethyl-3-(4-sulfonatobutyl)-1H,2H,3H-benzo[e]indol-2-ylidene]hepta-1,3,5-trien-1-yl]-1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-3-ium
Molecular FormulaC43H47N2NaO6S2
InChI KeyMOFVSTNWEDAEEK-UHFFFAOYSA-M
SMILES[Na+].CC1(C)C(=C/C=C/C=CC=CC2=[N+](CCCCS([O-])(=O)=O)C3=CC=C4C=CC=CC4=C3C2(C)C)N(CCCCS([O-])(=O)=O)C2=CC=C3C=CC=CC3=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV Lambda max784 nm (in Methanol)
Molar absorption coefficient>=230000 l/(mol.cm)
Appearance (Form)Crystalline powder or crystals
Appearance (Color)Green to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only