2-Methyl-1-propanol, for HPLC
2-Methyl-1-propanol, for HPLC
2-Methyl-1-propanol, for HPLC
Thermo Scientific Chemicals

2-Methyl-1-propanol, for HPLC

Green features
CAS: 78-83-1 | C4H10O | 74.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4126400101 LGlass bottle
Catalog number 412640010
Price (USD)
149.65
Special offer
Online exclusive
Ends: 28-Dec-2026
176.00
Save 26.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
149.65
Special offer
Online exclusive
Ends: 28-Dec-2026
176.00
Save 26.35
Each
Chemical Identifiers
CAS78-83-1
IUPAC Name2-methylpropan-1-ol
Molecular FormulaC4H10O
InChI KeyZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILESCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.45 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Color scale=<10 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only