Isobutyl methacrylate, 99%, stabilized
Isobutyl methacrylate, 99%, stabilized
Isobutyl methacrylate, 99%, stabilized
Thermo Scientific Chemicals

Isobutyl methacrylate, 99%, stabilized

CAS: 97-86-9 | C8H14O2 | 142.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 412680025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS97-86-9
IUPAC Name2-methylpropyl 2-methylprop-2-enoate
Molecular FormulaC8H14O2
InChI KeyRUMACXVDVNRZJZ-UHFFFAOYSA-N
SMILESCC(C)COC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Acidity=<0.005 % (as methacrylic acid)
Stabilizer15 to 20 ppm MEHQ
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only