3-Methyl-1-butanol, ACS reagent
3-Methyl-1-butanol, ACS reagent
3-Methyl-1-butanol, ACS reagent
Thermo Scientific Chemicals

3-Methyl-1-butanol, ACS reagent

CAS: 123-51-3 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 412730010
Price (USD)
136.65
Special offer
Online exclusive
Ends: 28-Dec-2026
161.00
Save 24.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 28-Dec-2026
161.00
Save 24.35
Each
Chemical Identifiers
CAS123-51-3
IUPAC Name3-methylbutan-1-ol
Molecular FormulaC5H12O
InChI KeyPHTQWCKDNZKARW-UHFFFAOYSA-N
SMILESCC(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Acids and esters=<0.2 % (as Amyl acetate)
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=98.5 %
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only