Isopropyl bromoacetate, 97%
Isopropyl bromoacetate, 97%
Isopropyl bromoacetate, 97%
Thermo Scientific Chemicals

Isopropyl bromoacetate, 97%

CAS: 29921-57-1 | C5H9BrO2 | 181.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 412811000
Price (USD)
221.00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
221.00
Each
Chemical Identifiers
CAS29921-57-1
IUPAC Namepropan-2-yl 2-bromoacetate
Molecular FormulaC5H9BrO2
InChI KeyJCWLEWKPXYZHGQ-UHFFFAOYSA-N
SMILESCC(C)OC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only