Lauric acid, sodium salt, 98%
Lauric acid, sodium salt, 98%
Lauric acid, sodium salt, 98%
Thermo Scientific Chemicals

Lauric acid, sodium salt, 98%

CAS: 629-25-4 | C12H23NaO2 | 222.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 413041000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS629-25-4
IUPAC Namesodium dodecanoate
Molecular FormulaC12H23NaO2
InChI KeyBTURAGWYSMTVOW-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Water=<3 %
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only