Linoleic acid, 70%
Linoleic acid, 70%
Linoleic acid, 70%
Thermo Scientific Chemicals

Linoleic acid, 70%

CAS: 60-33-3 | C18H32O2 | 280.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 413195000
Price (USD)
107.65
Special offer
Online exclusive
Ends: 28-Dec-2026
126.00
Save 18.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 28-Dec-2026
126.00
Save 18.35
Each
Chemical Identifiers
CAS60-33-3
IUPAC Name(9Z,12Z)-octadeca-9,12-dienoic acid
Molecular FormulaC18H32O2
InChI KeyOYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILESCCCCCC=C/CC=C/CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Acid value195 to 205 mg KOH/g
GC>=65 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only