Lithium chloride, 99+%, ACS reagent
Lithium chloride, 99+%, ACS reagent
Lithium chloride, 99+%, ACS reagent
Thermo Scientific Chemicals

Lithium chloride, 99+%, ACS reagent

CAS: 7447-41-8 | ClLi | 42.39 g/mol
数量:
5 g
500 g
2.5 kg
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 413270050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7447-41-8
IUPAC Namelithium(1+) chloride
Molecular FormulaClLi
InChI KeyKWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES[Li+].[Cl-]
さらに表示
Insoluble matter=<0.01 %
Barium (Ba)=<30 ppm
Potassium (K)=<100 ppm
Loss on drying=<1.0 % (1 g, 105°C, 2 hrs)
Titrable base=<0.008 meq/g
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

よくあるご質問(FAQ)

What storage conditions should I use for Lithium chloride?

Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.