What storage conditions should I use for Lithium chloride?
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.
Search
View additional product information for Lithium chloride, 99+%, ACS reagent - FAQs (413270025, 413271000, 413275000, 413270050)
1 product FAQs found