Lithium stearate, 95%
Lithium stearate, 95%
Lithium stearate, 95%
Thermo Scientific Chemicals

Lithium stearate, 95%

CAS: 4485-12-5 | C18H35LiO2 | 290.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 413340250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4485-12-5
IUPAC Namelithium(1+) octadecanoate
Molecular FormulaC18H35LiO2
InChI KeyHGPXWXLYXNVULB-UHFFFAOYSA-M
SMILES[Li+].CCCCCCCCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO4>=94 %
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only