Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm - FAQs
Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm - FAQs
View additional product information for Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm - FAQs (413380250)
1 product FAQs found
What are the dimensions of the Magnesium, Reagent, Ribbon, +99%?
The Magnesium, Reagent, Ribbon, +99% is one long ribbon with dimensions of 3.2-4.0 mm (width) x 0.3-0.4 mm (thickness).