4-Methoxybenzyl isothiocyanate, 95%
4-Methoxybenzyl isothiocyanate, 95%
4-Methoxybenzyl isothiocyanate, 95%
Thermo Scientific Chemicals

4-Methoxybenzyl isothiocyanate, 95%

CAS: 3694-57-3 | C9H9NOS | 179.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4138100505 gGlass bottle
Catalog number 413810050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3694-57-3
IUPAC Name1-(isothiocyanatomethyl)-4-methoxybenzene
Molecular FormulaC9H9NOS
InChI KeyIMFQYAJJXFXVMM-UHFFFAOYSA-N
SMILESCOC1=CC=C(CN=C=S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5925 to 1.5985 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear yellow
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only