Search
Search
| Número de catálogo | Tensión | Tecnología del sensor de CO2 | Control de oxígeno |
|---|---|---|---|
| 4141 | 230 V | Sensor de IR | Del 1 al 20 % |
| 4120 | 115 V | Sensor de IR | Ninguno |
| 4121TS también denominado 4121 | 230 V | Sensor de IR | Ninguno |
| 4140TS también denominado 4140 | 115 V | Sensor de IR | Del 1 al 20 % |
| 4110 | 115 V | Sensor de TC | Ninguno |
| 4111TS | 230 V | Sensor de TC | Ninguno |
| 4130TS | 115 V | Sensor de TC | Del 1 al 20 % |
| 4131 | 230 V | Sensor de TC | Del 1 al 20 % |
The Thermo Scientific™ Forma Series 3 Water Jacketed CO2 Incubator includes reliable water jacket technology with effortless Thermofisher Scientific™ iCAN™ touchscreen. Dependable temperature stability and HEPA clean room air purity are paired with the simplest way to control and monitor your incubator, ensuring protection for your important cell cultures, a combination that's truly hard to beat.
iCAN touchscreen interface
Temperature stability with Water Jacket design
Unique triple-wall construction delivers optimal temperature uniformity. Provides ultimate thermal stability for protection for your valuable cell cultures against ambient temperature swings and unexpected power outages.
Protection with HEPA air filtration
Minimize the risk of airborne contaminants entering the incubator from multiple door openings. In-chamber HEPA filtration reaches ISO 5 clean room air quality in 5 minutes by filtering the chamber air every 60 seconds.
Enhanced flexibility with complete O2 control
Many cell cultures thrive best in CO2 incubators with controlled levels of oxygen. Select an O2 option to simulate physiological or hypoxic environments in a range of 1 to 20%. A dedicated O2 display set point and control allow for accurate monitoring.