Methyl dimethoxyacetate, 98%
Methyl dimethoxyacetate, 98%
Methyl dimethoxyacetate, 98%
Thermo Scientific Chemicals

Methyl dimethoxyacetate, 98%

CAS: 89-91-8 | C5H10O4 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 414180050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS89-91-8
IUPAC Namemethyl 2,2-dimethoxyacetate
Molecular FormulaC5H10O4
InChI KeyNZTCVGHPDWAALP-UHFFFAOYSA-N
SMILESCOC(OC)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4040 to 1.4070 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only