Methyl palmitate, 95%
Methyl palmitate, 95%
Methyl palmitate, 95%
Thermo Scientific Chemicals

Methyl palmitate, 95%

CAS: 112-39-0 | C17H34O2 | 270.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 414681000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-39-0
IUPAC Namemethyl hexadecanoate
Molecular FormulaC17H34O2
InChI KeyFLIACVVOZYBSBS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only