Methyl Red, sodium salt, ACS reagent
Methyl Red, sodium salt, ACS reagent
Methyl Red, sodium salt, ACS reagent
Thermo Scientific Chemicals

Methyl Red, sodium salt, ACS reagent

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 414840250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS845-10-3
IUPAC Namesodium 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular FormulaC15H14N3NaO2
InChI KeyGNTPCYMJCJNRQB-UHFFFAOYSA-M
SMILES[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
さらに表示
Appearance (Form)Powder
Infrared spectrumConforms
Clarity of solution(Aqueous) Passes test
Appearance (Color)Orange to red to brown
Clarity of alcohol solutionPasses test
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only