Methylsulfuric acid sodium salt, 99%, pract.
Methylsulfuric acid sodium salt, 99%, pract.
Methylsulfuric acid sodium salt, 99%, pract.
Thermo Scientific Chemicals

Methylsulfuric acid sodium salt, 99%, pract.

CAS: 512-42-5 | CH3NaO4S | 134.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 414901000
Price (USD)
372.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
372.00
Each
Chemical Identifiers
CAS512-42-5
IUPAC Namesodium methyl sulfate
Molecular FormulaCH3NaO4S
InChI KeyDZXBHDRHRFLQCJ-UHFFFAOYSA-M
SMILES[Na+].COS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only